Culture media preparator, MasterClave® 20
Catalog # 710-1962
Supplier: bioMérieux
Specifications
- Beskrivning:Masterclave® 20 with power supply
- Förp.:1
Specifications
About this item
The Masterclave® 20 is an automated preparation system for culture media, agars or broths, and is designed for used by qualified, trained laboratory professionals in the clinical, pharmaceutical, food processing and cosmetics industries. With its high core temperature precision control, and high efficiency stirring system, the Masterclave® 20 system optimises work flow and guarantees homogeneity, fertility and sterility.
- Capacity: 1 to 20 litres
- Productivity: removes the preparatory stages such as medium stirring and bottle autoclaving
- Reproducibility: avoids human error and ensures batch-to-batch reproducibility
- Flexibility: suitable for both agar and broth preparation
- Easy to use: simply add powder and distilled water to implement a cycle, no supervision required during the cycle
- Weight: 90 kgs
- W×D×H: 590×700×650
Different light indicators keep user informed about the state of the equipment. A Wi-Fi stick or Ethernet connection to access a network can be used to manage the data.
Delivery: The system includes 2 pressure relief filters, 3 sampling nozzles, 3 sampling nozzle spare gaskets, 2 drain plugs, 1 vessel cleaning brush, Water tubing 4 m, 2 tubing clips, water inlet filter with clips, User manual and power cable.