Neocarmine MS "Fesago" colour reagent, Sigma-Aldrich®
Catalog # 1.06731.1000
Supplier: Merck
CAS Number:
Specifications
- Förpackningstyp:Plastic bottle
- Förp.:1 L
- Seq:5037
Specifications
Specification Test Results
- Suitability for coloration of textiles:passes test
Specification Test Results
About this item
:
Density:1,01 g/cm3 (20 °C)
Storage Temperature:Omgivande