Vatten, Reagent Grade, meets analytical specification of USP, steril
Catalog # MDTC46-002-LF
Supplier: Corning
CAS Number:
Specifications
- Förpackningstyp:Cube
- Förp.:2
- Seq:5178
- Pk Info:2× 4 l
Specifications
About this item
:
:18.02 g/mol
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332