Sealed Cuvette
Katalog # PERCL1270954
Leverantör: PerkinElmer
Specifikationer
- Skiktdjup:0,1 mm
- Beskrivning:Rectangular sealed liquid cell
- Förp.:1
Specifikationer
Om denna produkt
Rectangular cuvette filled by potassium bromide (KBr) for a wider spectral range than sodium chloride (NaCl) but more hygroscopic.