Orders LinkContactUsLinkComponent
Single-point sampler
Single-point sampler
Supplier:   Sampling Systems
Single-point sampler
Supplier:  Sampling Systems
Certificates
Single-point sampler
Single-point sampler
2 Options Available Below
Product Details & Documents
Single-point sampler for ice, wax, and similar materials.
Product Family Options
Product Information
  • Beskrivning
    Material
    Längd
    Ø
    Volym
    Availability
    Price
  • Ice sampler
    Rostfritt stål 316L
    550 mm
    40 mm
    50 ml
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 6 075,00 kr
    6 075,00 kr
    1 * 1 Items
       6 075,00 kr
  • Ice sampler
    Rostfritt stål 316L
    1050 mm
    40 mm
    50 ml
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 6 625,00 kr
    6 625,00 kr
    1 * 1 Items
       6 625,00 kr