Detergents for glassware washers, ProCare Lab 10 AT
Leverantör: MIELE
Certifikat
Om denna produkt
Liquid, alkaline detergent for machine based cleaning of laboratory glassware and laboratory utensils.
- Removes stubborn residues such as oils, greases, paraffins, resins and organic dyes
- Free from phosphates, silicates and oxidative agents
ProCare Lab 10 AT is intended for food-processing, cosmetics and pharmaceutical industries as well as the mineral oil sector and other industries.
Note: Suitable for stainless steel, laboratory glassware, ceramics, plastics such as PE, PP, PVDF, PTFE. Not suitable for use on non ferrous metals, light alloys, anodised aluminium, and aluminium.