Acidic cleaner and neutraliser, ProCare Lab 30 P
Catalog # MIEL10266490
Supplier: MIELE
Specifications
- Kapacitet:5 l
- Beskrivning:Acidic cleaner, ProCare lab 30 P
- Seq:01
- Förp.:5 L
- pH-värde:1,9 - 2,2
Specifications
About this item
Liquid, acidic cleaning concentrate for automated cleaning of laboratory glassware and reusable laboratory material.
- Good rinsing properties
- Free from tensides
- Removes limescale and other soils soluble in acids
ProCare Lab 30 P is intended for general use in medical, biological, microbiological and chemical laboratories, food processing, cosmetics and pharmaceutical industries as well as the mineral oil sector and other branches.
Note: Suitable for stainless steel, laboratory glassware, plastics such as PE, PP, PVDF, PTFE. Not suitable for use on non ferrous metals, light alloys, anodised aluminium, aluminium, chrome- and nickel-plated parts.