ContactUsLinkComponent
Epidural anaesthesia needle, Perican®
Epidural anaesthesia needle, Perican®
Katalog # BRAU4512383
Leverantör:  B. Braun
undefined
Epidural anaesthesia needle, Perican®
Katalog # BRAU4512383
Leverantör:  B. Braun
Specifikationer
  • Mätare:
    G 18
  • Färg:
    Rosa
  • Beskrivning:
    Epidural anaesthesia needle with Tuohy bevel
  • Längd:
    83 mm
  • Förp.:
    100
Specifikationer
Om denna produkt
Epidural anaesthesia needle with Tuohy-type bevel.
  • Design for different tissue layers
  • Control of needle advancement and small wings
  • Transparent needle with colour-coded stylet hub
Suitable for all procedures specially for epidural anesthesia.