Examination gloves, nitrile, Sempercare® nitrile shine+
Supplier: SEMPERIT TECHN PROD
Certificates
About this item
Disposable nitrile gloves with textured fingertips and longer cuff.
- Greater wall thickness in order to prevent potential tears
- New foil opening: Easier to remove
- Textured fingertips for improved grip
The longer cuff makes these gloves ideal for use in critical applications especially in the nursing sector.
Certifications: EN 455, EN 374-1/2-/3; Cat. III (PPE); Cat I (MD); AQL 1,5.
Specifications
- Catalog No:
- SEMP826778433
- SEMP826778435
- SEMP826778437
- Ej handspecifika/handspecifika:
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Engångs/Återanvändbar:
- Engångs
- Engångs
- Engångs
- Latex/latexfri:
- Latexfria
- Latexfria
- Latexfria
- Pudrad/puderfri:
- Puderfria
- Puderfria
- Puderfria
- Yta:
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Kragmodell:
- Med rullad kant
- Med rullad kant
- Med rullad kant
- EN standard:
- EN 455, EN 374-1/2-/3
- EN 455, EN 374-1/2-/3
- EN 455, EN 374-1/2-/3