Cut resistant gloves, HyFlex® 11-541
Leverantör: Ansell
Certifikat
Om denna produkt
These seamless liners with INTERCEPT™ technology yarn feature a nitrile foam coating on palm and fingertips.
- Form-fitting ultralight weight glove
- ANSELL GRIP Technology™ minimises the force required to grip oily or wet tools or materials
- INTERCEPT™ Technology yarn has been designed to exceed industry standards
- Best-in-class cut protection and increased comfort with dexterity
- FORTIX™ Technology applies a thin, resilient and breathable nitrile foam coating, which extends gloves' working life and improves comfort in abrasive applications
Ideal for the assembly of small sharp parts, for the handling of small sharp metal parts, light duty stamping and maintenance.
Certifications: EN 388:2016 (4X21D), EN 407 (X1XXXX); Cat. III.
Specifikationer
- Catalog No:
- ANSE11-541/11
- ANSE11-541/7
- ANSE11-541/6
- ANSE11-541/9
- ANSE11-541/10
- ANSE11-541/8
- Modell:
- 11-541
- 11-541
- 11-541
- 11-541
- 11-541
- 11-541
- Ej handspecifika/handspecifika:
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Engångs/Återanvändbar:
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Fodermaterial:
- PE
- PE
- PE
- PE
- PE
- PE
- Yta:
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Mätare:
- 18
- 18
- 18
- 18
- 18
- 18
- Kragmodell:
- Stickad mudd
- Stickad mudd
- Stickad mudd
- Stickad mudd
- Stickad mudd
- Stickad mudd
- EN standard:
- EN 388:2016 (4X21D), EN 407 (X1XXXX)
- EN 388:2016 (4X21D), EN 407 (X1XXXX)
- EN 388:2016 (4X21D), EN 407 (X1XXXX)
- EN 388:2016 (4X21D), EN 407 (X1XXXX)
- EN 388:2016 (4X21D), EN 407 (X1XXXX)
- EN 388:2016 (4X21D), EN 407 (X1XXXX)