Order Entry
Sweden
ContactUsLinkComponent
Anti-MyD88 Rabbit Polyclonal Antibody
Anti-MyD88 Rabbit Polyclonal Antibody
Katalog # ENZOADICSA510E
Leverantör:  ENZO LIFE SCIENCES
Anti-MyD88 Rabbit Polyclonal Antibody
Katalog # ENZOADICSA510E
Leverantör:  ENZO LIFE SCIENCES

Specifikationer

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Myeloid differentiation primary response 88
  • Antigensymbol:
    MYD88
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid
  • Antigen synonymer:
    MYD88D|myeloid differentiation primary response 88
  • Storage buffer:
    PBS containing 0,02% sodium azide
  • Förvaringstemperatur:
    4 °C
  • Shipping temperature:
    blue ice not frozen
  • Immunogen:
    Synthetic peptide corresponding to a portion of human MyD88. The sequence is completely conserved in mouse
  • Rening:
    Purified by peptide affinity chromatography
  • Storlek:
    100 μg
  • Förp.:
    100 µG

Specifikationer

Om denna produkt

MyD88 (Myeloid differentiation primary response gene 88) is a general adapter protein for the Toll/IL-1R family of receptors that have been shown to induce growth arrest, terminal differentiation, and apoptosis.

  • Dilution Range: 1-2 µg/ml, ECL for Western blot
  • Detects a band of ~35kDa by Western blot

MyD88 plays an important role in the inflammatory response induced by cytokines, IL-1 and IL-18, and endotoxin. It has been shown that signaling via MyD88 is essential for an LPS response. MyD88 is a cytoplasmic protein, which is expressed in many tissues.

Type: Primary
Antigen: Myeloid differentiation primary response gene 88
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat