Anti-VIM Mouse Monoclonal Antibody () [clone: V9]
Katalog # DIABPDM029
Leverantör: DIAGNOSTIC BIOSYSTEM
Specifikationer
- Antikroppsnamn:Vimentin
- Antigensymbol:VIM
- Klonalitet:Monoclonal
- Klon:V9
- Värd:Mouse
- Isotyp:IgG1 kappa
- Antigen synonymer:CTRCT30|HEL113
- Storlek:6 mL
- Förp.:6 ml
Specifikationer
Om denna produkt
Type:
Antigen: VIM
Clonality: Monoclonal
Clone: V9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: