Anti-VIM Rabbit Polyclonal Antibody
Leverantör: DIAGNOSTIC BIOSYSTEM
Certifikat
Om denna produkt
Type:
Antigen: VIM
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifikationer
- Catalog No:
- DIABRP115-05
- DIABRP115
- Antigensymbol:
- VIM
- VIM
- Antikroppsnamn:
- Vimentin
- Vimentin
- Antigen synonymer:
- CTRCT30|HEL113
- CTRCT30|HEL113
- Klonalitet:
- Polyclonal
- Polyclonal
- Värd:
- Rabbit
- Rabbit