Anti-VIM Mouse Monoclonal Antibody () [clone: V9]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: VIM
Clonality: Monoclonal
Clone: V9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Specifications
- Catalog No:
- DIABMOB090-05
- DIABMOB090
- Antigensymbol:
- VIM
- VIM
- Antikroppsnamn:
- Vimentin
- Vimentin
- Antigen synonymer:
- CTRCT30|HEL113
- CTRCT30|HEL113
- Klonalitet:
- Monoclonal
- Monoclonal
- Klon:
- V9
- V9
- Värd:
- Mouse
- Mouse
- Isotyp:
- IgG1 kappa
- IgG1 kappa