Anti-RAS Rabbit Polyclonal Antibody
Leverantör: DIAGNOSTIC BIOSYSTEM
Certifikat
Om denna produkt
Type:
Antigen: RAS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: 0
Specifikationer
- Catalog No:
- DIABRP094-05
- DIABRP094
- Antigensymbol:
- RAS
- RAS
- Klonalitet:
- Polyclonal
- Polyclonal
- Värd:
- Rabbit
- Rabbit