Anti-CDKN1B Rabbit Polyclonal Antibody
Leverantör: DIAGNOSTIC BIOSYSTEM
Certifikat
Om denna produkt
Type:
Antigen: CDKN1B
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifikationer
- Catalog No:
- DIABRP102
- DIABRP102-05
- Antigensymbol:
- CDKN1B
- CDKN1B
- Antikroppsnamn:
- Cyclin-dependent kinase inhibitor 1B
- Cyclin-dependent kinase inhibitor 1B
- Antigen synonymer:
- CDKN4|KIP1|MEN1B|P27KIP1 |MEN4 |p27Kip1
- CDKN4|KIP1|MEN1B|P27KIP1 |MEN4 |p27Kip1
- Klonalitet:
- Polyclonal
- Polyclonal
- Värd:
- Rabbit
- Rabbit