Anti-P501S Mouse Monoclonal Antibody [clone: 10000]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: P501S
Clonality: Monoclonal
Clone: 10000
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: 0
Specifications
- Catalog No:
- DIABMOB439-05
- DIABMOB439
- Antigensymbol:
- P501S
- p501S
- Klonalitet:
- Monoclonal
- Monoclonal
- Klon:
- 10E3
- 10E3
- Värd:
- Mouse
- Mouse
- Isotyp:
- IgG2a
- IgG2a