Anti-Myosin Mouse Monoclonal Antibody [clone: MY32]
Catalog # DIABPDM085
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antikroppsnamn:Myosin
- Antigensymbol:MYHSA1
- Klonalitet:Monoclonal
- Klon:MY32
- Värd:Mouse
- Isotyp:IgG1
- Antigen synonymer:MGC133384|MYHa|MYHSA1|MyHC-2X/D
- Storlek:6 mL
- Förp.:6 ml
Specifications
About this item
Type:
Antigen: Myosin
Clonality: Monoclonal
Clone: MY32
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: