Anti-p14ARF Rabbit Polyclonal Antibody
Leverantör: DIAGNOSTIC BIOSYSTEM
Certifikat
Om denna produkt
Type:
Antigen: p14ARF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: 0
Specifikationer
- Catalog No:
- DIABRP084-05
- DIABRP084
- Antigensymbol:
- p14ARF
- p14ARF
- Antikroppsnamn:
- Cyclin-dependent kinase inhibitor 2A (melanoma p16 inhibits CDK4)
- Cyclin-dependent kinase inhibitor 2A (melanoma p16 inhibits CDK4)
- Antigen synonymer:
- ARF, CDK4I, CDKN2, CMM2, INK4, INK4a, MLM, MTS1, TP16, p14, p14ARF, p16, p16INK4, p16INK4a, p19, p19Arf
- ARF, CDK4I, CDKN2, CMM2, INK4, INK4a, MLM, MTS1, TP16, p14, p14ARF, p16, p16INK4, p16INK4a, p19, p19Arf
- Klonalitet:
- Polyclonal
- Polyclonal
- Värd:
- Rabbit
- Rabbit