Anti-MPO Rabbit Polyclonal Antibody
Leverantör: DIAGNOSTIC BIOSYSTEM
Certifikat
Om denna produkt
Type:
Antigen: MPO
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifikationer
- Catalog No:
- DIABRP053
- DIABRP053-05
- Antigensymbol:
- MPO
- MPO
- Antikroppsnamn:
- Myeloperoxidase
- Myeloperoxidase
- Antigen synonymer:
- Myelin Membrane Encephalitogenic Protei|Myelin A1 Protein
- Klonalitet:
- Polyclonal
- Polyclonal
- Värd:
- Rabbit
- Rabbit