Anti-ENO2 Mouse Monoclonal Antibody [clone: VI-H14]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: ENO2
Clonality: Monoclonal
Clone: VI-H14
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Specifications
- Catalog No:
- DIABMOB212
- DIABMOB212-05
- Antigensymbol:
- ENO2
- NSE
- Antikroppsnamn:
- Enolase 2
- Neuron Specific Enolase
- Antigen synonymer:
- Gamma-enolase|Enolase 2|ENO2|Neural enolase|NSE|2-phospho-D-glycerate hydro-lyase|Neuron-specific enolase
- Eno|Neuron Specific Enolase|ENOG|Neuronal Enolase|Neural Enolase|glycerate hydrolyase
- Klonalitet:
- Monoclonal
- Monoclonal
- Klon:
- VI-H14
- VI-H14
- Värd:
- Mouse
- Mouse
- Isotyp:
- IgG1
- IgG1