Anti-Lysozyme Rabbit Polyclonal Antibody
Leverantör: DIAGNOSTIC BIOSYSTEM
Certifikat
Om denna produkt
Type:
Antigen: Lysozyme
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifikationer
- Catalog No:
- DIABRP028
- DIABRP028-05
- Antikroppsnamn:
- Lysozyme
- Lysozyme
- Antigen synonymer:
- lambda light chain|Ig lambda|Ig lambda light chain|Immunoglobulin lambda light chain|Igl
- lambda light chain|Ig lambda|Ig lambda light chain|Immunoglobulin lambda light chain|Igl
- Klonalitet:
- Polyclonal
- Polyclonal
- Värd:
- Rabbit
- Rabbit
- Epitop:
- Light Chain
- Light Chain