Anti-A2M Rabbit Polyclonal Antibody
Type:
Antigen: A2M
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
- Catalog No:
- DIABRP049
- DIABRP049-05
- Antigensymbol:
- A2M
- A2M
- Antikroppsnamn:
- Alpha-2-macroglobulin
- Alpha-2-macroglobulin
- Antigen synonymer:
- alpha-2-M|CPAMD5|C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5
- alpha-2-M|CPAMD5|C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5
- Klonalitet:
- Polyclonal
- Polyclonal
- Värd:
- Rabbit
- Rabbit