Anti-IgL Rabbit Polyclonal Antibody
Catalog # DIABPDR016
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antikroppsnamn:Immunoglobulin L
- Antigensymbol:IgL
- Klonalitet:Polyclonal
- Värd:Rabbit
- Epitop:Light Chain
- Antigen synonymer:lambda light chain|Ig lambda|Ig lambda light chain|Immunoglobulin lambda light chain|Igl
- Storlek:6 mL
- Förp.:6 ml
Specifications
About this item
Type:
Antigen: IgL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: Light Chain
Host: Rabbit
Isotype:
Reactivity: