Anti-Insulin Mouse Monoclonal Antibody [clone: K36ac10]
Catalog # DIABMOB234
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antikroppsnamn:Insulin
- Klonalitet:Monoclonal
- Klon:K36ac10
- Värd:Mouse
- Isotyp:IgG1
- Storlek:1 mL
- Förp.:1 ml
Specifications
About this item
Type:
Antigen: Insulin
Clonality: Monoclonal
Clone: K36ac10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: