Anti-HSVII Mouse Monoclonal Antibody [clone: 12.3.4+1.1.1]
Catalog # DIABPDM081
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antikroppsnamn:Hepatitis B Virus Surface Antigen (ad/ay)
- Antigensymbol:HSVII
- Klonalitet:Monoclonal
- Klon:12.3.4+1.1.1
- Värd:Mouse
- Isotyp:IgG1 kappa,IgG1 lambda
- Antigen synonymer:HBsAg|Hepatitis B Surface Antigen|HepB|HepB ayw|HepB surface antigen|HBVgp2
- Storlek:6 mL
- Förp.:6 ml
Specifications
About this item
Type:
Antigen: HSVII
Clonality: Monoclonal
Clone: 12.3.4+1.1.1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 lambda + IgG1 kappa
Reactivity: