Anti-IgG Rabbit Polyclonal Antibody
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABRP023-05
- DIABRP023
- Antigensymbol:
- IgG
- IgG
- Antikroppsnamn:
- Immunoglobulin G
- Immunoglobulin G
- Antikroppstyp:
- Sekundär
- Sekundär
- Klonalitet:
- Polyclonal
- Polyclonal
- Värd:
- Rabbit
- Rabbit