Anti-PSAP Rabbit Polyclonal Antibody
Katalog # DIABPDR038
Leverantör: DIAGNOSTIC BIOSYSTEM
Specifikationer
- Antikroppsnamn:Prostate Specific Acid Phosphatase
- Antigensymbol:PSAP
- Klonalitet:Polyclonal
- Värd:Rabbit
- Antigen synonymer:SAP1|PAP|prostatic acid phosphotase|prostatic acid phosphatase|GLBA
- Storlek:6 mL
- Förp.:6 ml
Specifikationer
Om denna produkt
Type:
Antigen: PSAP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: 0