Anti-Cytokeratin Cocktail Mouse Monoclonal Antibody [clone: DBS/1]
DIABPDM038
- Antikroppsnamn:Cytokeratin Cocktail
- Antigensymbol:CK Cocktail
- Klonalitet:Monoclonal
- Klon:DBS/1
- Värd:Mouse
- Isotyp:IgG2a,IgG1
- Antigen synonymer:or transformation-related protein 53 (TRP53)|tumor suppressor p53|antigen NY-CO-13|P53|BCC7|TRP53|phosphoprotein p53|LFS1|tumor protein p53
- Storlek:6 mL
- Förp.:6 ml
Type:
Antigen: Cytokeratin Cocktail
Clonality: Monoclonal
Clone: DBS/1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a + IgG1
Reactivity: