Anti-CK19 Rabbit Polyclonal Antibody
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: CK19
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABRP114-05
- DIABRP114
- Antigensymbol:
- CK19
- CK19
- Antikroppsnamn:
- Cytokeratin 19
- Cytokeratin 19
- Antigen synonymer:
- type I cytoskeletal 19|CK19|CK-19|K1CS|keratin-19|40-kd|K19|KRT19|cytokeratin-19|type I|keratin|40-kDa keratin intermediate filament
- type I cytoskeletal 19|CK19|CK-19|K1CS|keratin-19|40-kd|K19|KRT19|cytokeratin-19|type I|keratin|40-kDa keratin intermediate filament
- Klonalitet:
- Polyclonal
- Polyclonal
- Värd:
- Rabbit
- Rabbit