Anti-AACT Rabbit Polyclonal Antibody
Catalog # DIABPDR023
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antikroppsnamn:Serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3
- Antigensymbol:SERPINA3
- Klonalitet:Polyclonal
- Värd:Rabbit
- Antigen synonymer:MGC88254|ACT|GIG24|GIG25
- Storlek:6 mL
- Förp.:6 ml
Specifications
About this item
Type:
Antigen: AACT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: