Anti-HCG Rabbit Polyclonal Antibody
Leverantör: DIAGNOSTIC BIOSYSTEM
Certifikat
Om denna produkt
Type:
Antigen: hCG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifikationer
- Catalog No:
- DIABRP007-05
- DIABRP007
- Antigensymbol:
- HCG
- HCG
- Antikroppsnamn:
- Chorionic Gonadotrophin
- Chorionic Gonadotrophin
- Klonalitet:
- Polyclonal
- Polyclonal
- Värd:
- Rabbit
- Rabbit