Order Entry
Sweden
ContactUsLinkComponent
Anti-CD117/c-kit Rabbit Polyclonal Antibody
Anti-CD117/c-kit Rabbit Polyclonal Antibody
Katalog # DIABPDR045
Leverantör:  DIAGNOSTIC BIOSYSTEM
undefined
Anti-CD117/c-kit Rabbit Polyclonal Antibody
Katalog # DIABPDR045
Leverantör:  DIAGNOSTIC BIOSYSTEM

Specifikationer

  • Antikroppsnamn:
    KIT Proto-oncogene Receptor Tyrosine Kinase
  • Antigensymbol:
    KIT
  • Klonalitet:
    Polyclonal
  • Värd:
    Rabbit
  • Antigen synonymer:
    SCFR|CD117|PBT|C-Kit
  • Storlek:
    6 mL
  • Förp.:
    6 ml

Specifikationer

Om denna produkt

Type:
Antigen: CD117/c-kit
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: