Anti-NCAM1 Mouse Monoclonal Antibody [clone: 123C3.D5]
Catalog # DIABPDM110
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antikroppsnamn:Neural Cell Adhesion Molecule 1
- Antigensymbol:NCAM1
- Klonalitet:Monoclonal
- Klon:123C3.D5
- Värd:Mouse
- Isotyp:IgG1 kappa
- Antigen synonymer:NCAM|MSK39|CD56
- Storlek:6 mL
- Förp.:6 ml
Specifications
About this item
Type:
Antigen: NCAM1
Clonality: Monoclonal
Clone: 123C3.D5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: