Anti-Cd31 Endothelial Cell Mouse Monoclonal Antibody [clone: JC/70A]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: Cd31 Endothelial Cell
Clonality: Monoclonal
Clone: JC/70A
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Specifications
- Catalog No:
- DIABMOB034
- DIABMOB034-05
- Antigensymbol:
- CD31
- Cd31 Endothelial Cell
- Antikroppsnamn:
- CD31 Molecule
- Antigen synonymer:
- endoCAM|PECAM-1|GPIIA'|EndoCAM|PECAM1|adhesion molecule|CD31|PECA1
- Klonalitet:
- Monoclonal
- Monoclonal
- Klon:
- JC/70A
- JC/70A
- Värd:
- Mouse
- Mouse
- Isotyp:
- IgG1 kappa
- IgG1 kappa