Anti-CD23 Mouse Monoclonal Antibody [clone: 1B12]
Leverantör: DIAGNOSTIC BIOSYSTEM
Certifikat
Om denna produkt
Type:
Antigen: CD23
Clonality: Monoclonal
Clone: 1B12
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Specifikationer
- Catalog No:
- DIABMOB294
- DIABMOB294-05
- Antigensymbol:
- FCE2
- FCE2
- Antikroppsnamn:
- Fc fragment of IgE receptor 2
- Fc fragment of IgE receptor 2
- Antigen synonymer:
- CLEC4J|BLAST-2|FCE2|CD23A|CD23|IGEBF
- CLEC4J|BLAST-2|FCE2|CD23A|CD23|IGEBF
- Klonalitet:
- Monoclonal
- Monoclonal
- Klon:
- 1B12
- 1B12
- Värd:
- Mouse
- Mouse
- Isotyp:
- IgG1 kappa
- IgG1 kappa