Anti-CD8 Mouse Monoclonal Antibody [clone: 144B]
DIABPDM094
- Antikroppsnamn:Cluster of differentiation 8
- Antigensymbol:CD8
- Klonalitet:Monoclonal
- Klon:144B
- Värd:Mouse
- Isotyp:IgG1 kappa
- Antigen synonymer:CD8alpha|p32|Leu-2|CD8a|MAL|CD8|CD8b|CD8beta
- Storlek:6 mL
- Förp.:6 ml
Type:
Antigen: CD8
Clonality: Monoclonal
Clone: 144B
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: