Anti-CD3 (T Cell) Rabbit Monoclonal Antibody
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: CD3 (T Cell)
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABRP005
- DIABRP005-05
- Antigensymbol:
- CD3 (T Cell)
- CD3
- Antikroppsnamn:
- CD3 Molecule
- Klonalitet:
- Monoclonal
- Monoclonal
- Värd:
- Rabbit
- Rabbit