Anti-CASP8 Rabbit Polyclonal Antibody
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: CASP8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABRP097
- DIABRP097-05
- Antigensymbol:
- CASP8
- CASP8
- Antikroppsnamn:
- Caspase 8
- Caspase 8
- Antigen synonymer:
- FLICE|caspase 8|MACH-beta-1/2/3/4 protein|FADD-homologous ICE/CED-3-like protease|MCH5|apoptotic protease Mch-5|FADD-like ICE|MACH-alpha-1/2/3 protein|CAP4|ALPS2B|apoptotic cysteine protease|Casp-8|MORT1-associated ced-3 homolog|caspase-8|apoptosis-related cysteine protease|ICE-like apoptotic protease 5|MACH
- FLICE|caspase 8|MACH-beta-1/2/3/4 protein|FADD-homologous ICE/CED-3-like protease|MCH5|apoptotic protease Mch-5|FADD-like ICE|MACH-alpha-1/2/3 protein|CAP4|ALPS2B|apoptotic cysteine protease|Casp-8|MORT1-associated ced-3 homolog|caspase-8|apoptosis-related cysteine protease|ICE-like apoptotic protease 5|MACH
- Klonalitet:
- Polyclonal
- Polyclonal
- Värd:
- Rabbit
- Rabbit