Anti-BCL6 Mouse Monoclonal Antibody () [clone: PG-B6P]
Leverantör: DIAGNOSTIC BIOSYSTEM
Certifikat
Om denna produkt
Type:
Antigen: BCL6
Clonality: Monoclonal
Clone: PG-B6P
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: 0
Specifikationer
- Catalog No:
- DIABMOB396
- DIABMOB396-05
- Antigensymbol:
- BCL6
- BCL6
- Antikroppsnamn:
- B-cell lymphoma 6 protein
- B-cell lymphoma 6 protein
- Antigen synonymer:
- BCL5|LAZ3|BCL6A|ZNF51|ZBTB27
- BCL5|LAZ3|BCL6A|ZNF51|ZBTB27
- Klonalitet:
- Monoclonal
- Monoclonal
- Klon:
- PG-B6P
- PG-B6P
- Värd:
- Mouse
- Mouse
- Isotyp:
- IgG1 kappa
- IgG1 kappa