Anti-ACTH Rabbit Polyclonal Antibody
Katalog # DIABRP045
Leverantör: DIAGNOSTIC BIOSYSTEM
Specifikationer
- Antikroppsnamn:Adrenocorticotrophic Hormone
- Antigensymbol:ACTH
- Klonalitet:Polyclonal
- Värd:Rabbit
- Antigen synonymer:MSH|NPP|beta-LPH|alpha-melanocyte-stimulating hormone|adrenocorticotropic hormone|lipotr|POC|beta-melanocyte-stimulating hormone|gamma-MSH|alpha-MSH|beta-endorphin|CLIP|LPH|Corticotropin-lipotropin|adrenoCorticotropin|POMC|gamma-LPH|Corticotropin-like intermediary peptide|ACTH|beta-MSH
- Storlek:1 mL
- Förp.:1 ml
Specifikationer
Om denna produkt
Type:
Antigen: ACTH
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: