Anti-AFP Rabbit Polyclonal Antibody
DIABPDR013
- Antikroppsnamn:Alpha 1 Fetoprotein Receptor
- Antigensymbol:AFP
- Klonalitet:Polyclonal
- Värd:Rabbit
- Antigen synonymer:FETA|Alpha-1-Fetoprotein|HPAFP
- Storlek:6 mL
- Förp.:6 ml
Type:
Antigen: AFP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: