Anti-FLT4 Rabbit Polyclonal Antibody
Type:
Antigen: Flt4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
- Catalog No:
- DIABRP135-05
- DIABRP135
- Antigensymbol:
- FLT4
- FLT4
- Antikroppsnamn:
- FMS Related Tyrosine Kinase 4
- FMS Related Tyrosine Kinase 4
- Antigen synonymer:
- VEGFR3|LMPH1A|VEGFR-3|VEGF Receptor 3|fms related tyrosine kinase 4|FLT41|FLT-4|Vascular endothelial growth factor receptor 3|PCL
- VEGFR3|LMPH1A|VEGFR-3|VEGF Receptor 3|fms related tyrosine kinase 4|FLT41|FLT-4|Vascular endothelial growth factor receptor 3|PCL
- Klonalitet:
- Polyclonal
- Polyclonal
- Värd:
- Rabbit
- Rabbit