Anti-GCDFP15 Mouse Monoclonal Antibody [clone: 23A3]
Leverantör: DIAGNOSTIC BIOSYSTEM
Certifikat
Om denna produkt
Type:
Antigen: GCDFP15
Clonality: Monoclonal
Clone: 23A3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Specifikationer
- Catalog No:
- DIABMOB307-05
- DIABMOB307
- Antigensymbol:
- GCDFP15
- GCDFP15
- Antikroppsnamn:
- Gross Cystic Disease Fluid Protein 15
- Gross Cystic Disease Fluid Protein 15
- Antigen synonymer:
- GCDGP-15|SABP|Gross cystic disease fluid protein 15|Gp17|GPIP4|Prolactin induced protein|Prolactin inducible protein precursor|Secretory actin binding protein.|GCDGP 15
- GCDGP-15|SABP|Gross cystic disease fluid protein 15|Gp17|GPIP4|Prolactin induced protein|Prolactin inducible protein precursor|Secretory actin binding protein.|GCDGP 15
- Klonalitet:
- Monoclonal
- Monoclonal
- Klon:
- 23A3
- 23A3
- Värd:
- Mouse
- Mouse
- Isotyp:
- IgG2a kappa
- IgG2a kappa