Anti-TIMP4 Rabbit Polyclonal Antibody
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: TIMP4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABRP073-05
- DIABRP073
- Antigensymbol:
- TIMP4
- TIMP4
- Antikroppsnamn:
- Tissue Inhibitor of Metalloproteinase 4
- Tissue Inhibitor of Metalloproteinase 4
- Antigen synonymer:
- metalloproteinase inhibitor 4|tissue inhibitor of metalloproteinase 4|tissue inhibitor of metalloproteinases 4
- metalloproteinase inhibitor 4|tissue inhibitor of metalloproteinase 4|tissue inhibitor of metalloproteinases 4
- Klonalitet:
- Polyclonal
- Polyclonal
- Värd:
- Rabbit
- Rabbit