Anti-Elastin Mouse Monoclonal Antibody [clone: BA-4]
Leverantör: DIAGNOSTIC BIOSYSTEM
Certifikat
Om denna produkt
This antibody recognizes insoluble elastin, α-elastin, soluble non-cross linked precursor of elastin (tropoelastin). Elastin is an important polymeric protein of connective tissue that imparts elasticity to vertebrate elastic tissues.
Type:
Antigen: Elastin
Clonality: Monoclonal
Clone: BA-4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Specifikationer
- Catalog No:
- DIABMOB230-05
- DIABMOB230
- Antikroppsnamn:
- Elastin
- Elastin
- Klonalitet:
- Monoclonal
- Monoclonal
- Klon:
- BA-4
- BA-4
- Värd:
- Mouse
- Mouse
- Isotyp:
- IgG1
- IgG1