Anti-CK20 Mouse Monoclonal Antibody [clone: KS 20.8]
Leverantör: DIAGNOSTIC BIOSYSTEM
Certifikat
Om denna produkt
Cytokeratin 20 is less acidic than the other type I cytokeratins and is of particular interest because of its restricted range of expression. This antibody reacts with cytokeratin polypeptide of 46 kDa in immunoblot analyses. Studies have shown that a marked difference exists in the expression of Cytokeratin 20 among various carcinomas.
Type:
Antigen: CK20
Clonality: Monoclonal
Clone: KS 20.8
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Specifikationer
- Catalog No:
- DIABMOB123-05
- DIABMOB123
- DIABPDM049
- Antigensymbol:
- CK20
- CK20
- CK20
- Antikroppsnamn:
- Cytokeratin 20
- Cytokeratin 20
- Cytokeratin 20
- Antigen synonymer:
- keratin-20|Cytokeratin-20|protein IT|type I cytoskeletal 20|keratin
- keratin-20|Cytokeratin-20|protein IT|type I cytoskeletal 20|keratin
- keratin-20|Cytokeratin-20|protein IT|type I cytoskeletal 20|keratin
- Klonalitet:
- Monoclonal
- Monoclonal
- Monoclonal
- Klon:
- KS 20.8
- KS 20.8
- KS 20.8
- Värd:
- Mouse
- Mouse
- Mouse
- Isotyp:
- IgG2a kappa
- IgG2a kappa
- IgG2a kappa