Anti-CK19 Mouse Monoclonal Antibody [clone: BA17]
This antibody recognizes keratin polypeptide of 40 kDa. It is the smallest human keratin found in most simple and non-keratinizing stratified epithelia. This antigen is not present in adult intrafollicular epidermis. The antibody stains most of the epithelial cell types including ductal and glandular epithelia.
Type:
Antigen: CK19
Clonality: Monoclonal
Clone: BA17
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
- Catalog No:
- DIABMOB058-05
- DIABMOB058
- Antigensymbol:
- CK19
- CK19
- Antikroppsnamn:
- Cytokeratin 19
- Cytokeratin 19
- Antigen synonymer:
- type I cytoskeletal 19|CK19|CK-19|K1CS|keratin-19|40-kd|K19|KRT19|cytokeratin-19|type I|keratin|40-kDa keratin intermediate filament
- type I cytoskeletal 19|CK19|CK-19|K1CS|keratin-19|40-kd|K19|KRT19|cytokeratin-19|type I|keratin|40-kDa keratin intermediate filament
- Klonalitet:
- Monoclonal
- Monoclonal
- Klon:
- BA17
- BA17
- Värd:
- Mouse
- Mouse
- Isotyp:
- IgG1 kappa
- IgG1 kappa