Anti-C-Erbb-2 Rabbit Polyclonal Antibody
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
This antibody reacts with the c-erbB-2 oncoprotein, a 190 kDa protein. The proto-oncogene for c-erbB-2 is located on the human chromosome 17, band 21. It has structural similarities to EGFR. Several studies have indicated that c-erbB-2 may be a good indicator of the prognosis of human carcinomas in the breast, ovary, uterus and gastrointestinal tract.
Type:
Antigen: C-Erbb-2
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABRP006-05
- DIABRP006
- DIABPDR003
- Antigensymbol:
- C-ERBB2
- C-ERBB2
- C-ERBB2
- Antikroppsnamn:
- V-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2
- V-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2
- V-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2
- Klonalitet:
- Polyclonal
- Polyclonal
- Polyclonal
- Värd:
- Rabbit
- Rabbit
- Rabbit