Anti-IgG3 Mouse Monoclonal Antibody [clone: HBME-1]
DIABPDM138
- Antikroppsnamn:Immunoglobulin G3
- Antigensymbol:IgG3
- Klonalitet:Monoclonal
- Klon:HBME-1
- Värd:Mouse
- ImmunKemi:Yes
- Isotyp:IgM
- Reaktivitet:Human
- Storlek:6 mL
- Förp.:6 ml
Type:
Antigen: IgG3
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human